siRNA Details
| Gene Symbol: Zbtb8os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025970.3 | NP_080246.1 | 3 | 175 | ||
| GenBank mRNA | AK007377.1 | - | 3 | 165 | ||
| AK008007.1 | - | 3 | 176 | |||
| AK132279.1 | - | 186 | ||||
| AY071852.1 | - | 3 | 164 | |||
| BC026442.1 | - | 170 | ||||
| BC037657.1 | - | 145 | ||||
| BC046825.1 | - | 182 | ||||
| BC094629.1 | - | 3 | 206 | 3376726 | ||
| BC125606.1 | - | 3 | 176 | 40130121 | ||
| BC125608.1 | - | 3 | 176 | 40130123 | ||

Zbtb8os
Genomic Map