siRNA Details
| Gene Symbol: Trappc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093759.1 | NP_001087228.1 | 6 | 774 | ||
| NM_001093760.1 | NP_001087229.1 | 6 | 774 | |||
| NM_025879.2 | NP_080155.3 | 6 | 774 | |||
| GenBank mRNA | AK010346.1 | - | 6 | 755 | ||
| AK013956.1 | - | 6 | 732 | |||
| AK084373.1 | - | 6 | 724 | |||
| AK149832.1 | - | 6 | 777 | |||
| AK163342.1 | - | 6 | 762 | |||
| AK167749.1 | - | 6 | 734 | |||
| BC021756.1 | - | 6 | 715 | 3584010 | ||

Trappc13
Genomic Map