siRNA Details
| Gene Symbol: Pole4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025882.3 | NP_080158.1 | 2 | 302 | ||
| GenBank mRNA | AK010135.1 | - | 2 | 297 | ||
| AK010300.1 | - | 2 | 303 | |||
| AK011595.1 | - | 2 | 314 | |||
| AK017956.1 | - | 2 | 286 | |||
| AK145365.1 | - | 2 | 308 | |||
| AK160582.1 | - | 2 | 314 | |||
| AK167631.1 | - | 2 | 292 | |||
| AK182530.1 | - | 1 | 62 | |||
| BC023189.1 | - | 2 | 280 | 4952949 | ||

Pole4
Genomic Map