siRNA Details
| Gene Symbol: Zfp830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025884.4 | NP_080160.2 | 1 | 521 | ||
| GenBank mRNA | AK008475.1 | - | 1 | 521 | ||
| AK010353.1 | - | 1 | 512 | |||
| AK017640.1 | - | 1 | 501 | |||
| AK050043.1 | - | 1 | 516 | |||
| AK132497.1 | - | 1 | 522 | |||
| AK148058.1 | - | 1 | 522 | |||
| AK167656.1 | - | 1 | 512 | |||
| AK168417.1 | - | 1 | 501 | |||
| BC024340.1 | - | 1 | 507 | 4981533 | ||

Zfp830
Genomic Map