siRNA Details
| Gene Symbol: Tmem88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025915.4 | NP_080191.3 | 2 | 545 | ||
| GenBank mRNA | AK010789.1 | - | 2 | 552 | ||
| AK011240.1 | - | 2 | 546 | |||
| AK045149.1 | - | 2 | 546 | |||
| AK161789.1 | - | 2 | 546 | |||
| BC100316.1 | - | 2 | 534 | |||
| BC120912.2 | - | 2 | 546 | 40091214 | ||

Tmem88
Genomic Map