siRNA Details
| Gene Symbol: Mrpl45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025927.4 | NP_080203.1 | 2 | 348 | ||
| GenBank mRNA | AK011163.1 | - | 2 | 217 | ||
| AK011605.1 | - | 2 | 207 | |||
| AK081922.1 | - | 2 | 209 | |||
| AK157771.1 | - | 2 | 210 | |||
| BC020130.1 | - | 2 | 193 | 3990098 | ||
| BY263659.1 | - | 2 | 348 | |||

Mrpl45
Genomic Map