siRNA Details
| Gene Symbol: Syap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025932.2 | NP_080208.2 | 3 | 566 | ||
| GenBank mRNA | AK008390.1 | - | 3 | 535 | ||
| AK014893.1 | - | 3 | 552 | |||
| AK051251.1 | - | 3 | 523 | |||
| AK147055.1 | - | 3 | 502 | |||
| AK161413.1 | - | 3 | 523 | |||
| BC021373.1 | - | 3 | 496 | 5039534 | ||

Syap1
Genomic Map