siRNA Details
| Gene Symbol: Dynlrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291108.1 | NP_001278037.1 | 4 | 770 | ||
| NM_025947.3 | NP_080223.2 | 4 | 526 | |||
| GenBank mRNA | AK002359.1 | - | 4 | 467 | ||
| AK003435.1 | - | 3 | 456 | |||
| AK008595.1 | - | 5 | 488 | |||
| AK166685.1 | - | 4 | 502 | |||
| BC099423.1 | - | 4 | 486 | 30933448 | ||
| BC125610.1 | - | 4 | 486 | 40130125 | ||
| BC125648.1 | - | 4 | 486 | 40130163 | ||

Dynlrb1
Genomic Map