siRNA Details
| Gene Symbol: Trappc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025960.3 | NP_080236.2 | 6 | 628 | ||
| GenBank mRNA | AK007965.1 | - | 6 | 627 | ||
| AK008022.1 | - | 6 | 628 | |||
| AK008462.2 | - | 6 | 627 | |||
| AK088832.1 | - | 6 | 627 | |||
| AK189448.1 | - | 2 | 186 | |||
| BC020128.1 | - | 5 | 577 | |||
| BC037154.1 | - | 6 | 665 | 2811950 | ||
| BC106114.1 | - | 6 | 526 | 4168174 | ||

Trappc6a
Genomic Map