siRNA Details
| Gene Symbol: Mmachc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025962.3 | NP_080238.2 | 3 | 448 | ||
| GenBank mRNA | AK007725.1 | - | 3 | 383 | ||
| AK012761.1 | - | 3 | 385 | |||
| AK084194.1 | - | 3 | 399 | |||
| AK213975.1 | - | 2 | 149 | |||
| BB614283.1 | - | 3 | 449 | |||
| BC054756.1 | - | 3 | 387 | 6411797 | ||

Mmachc
Genomic Map