siRNA Details
| Gene Symbol: Napa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025898.3 | NP_080174.1 | 11 | 2234 | ||
| GenBank mRNA | AK005372.1 | - | 11 | 2236 | ||
| AK078019.1 | - | 1 | 1292 | |||
| AK136689.1 | - | 11 | 2237 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Napa| Gene Symbol: Napa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025898.3 | NP_080174.1 | 11 | 2234 | ||
| GenBank mRNA | AK005372.1 | - | 11 | 2236 | ||
| AK078019.1 | - | 1 | 1292 | |||
| AK136689.1 | - | 11 | 2237 | |||