siRNA Details
| Gene Symbol: Dek | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025900.2 | NP_080176.2 | 4 | 456 | ||
| GenBank mRNA | AK012615.1 | - | 4 | 397 | ||
| AK012877.1 | - | 4 | 391 | |||
| AK033451.1 | - | 4 | 346 | |||
| AK154013.1 | - | 4 | 438 | |||
| BC055451.1 | - | 4 | 402 | 4164972 | ||

Dek
Genomic Map