siRNA Details
| Gene Symbol: 2010109I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025929.2 | NP_080205.1 | 3 | 880 | ||
| GenBank mRNA | AK008365.1 | - | 3 | 880 | ||
| AK018552.1 | - | 3 | 880 | |||
| AK038164.1 | - | 856 | ||||
| BC116397.1 | - | 3 | 860 | 40101946 | ||
| BC116398.1 | - | 3 | 860 | 40101947 | ||

2010109I03Rik
Genomic Map