siRNA Details
| Gene Symbol: Mmachc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025962.3 | NP_080238.2 | 4 | 1879 | ||
| GenBank mRNA | AK007725.1 | - | 4 | 1813 | ||
| AK084194.1 | - | 5 | 1875 | |||
| BC054756.1 | - | 4 | 1818 | 6411797 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmachc| Gene Symbol: Mmachc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025962.3 | NP_080238.2 | 4 | 1879 | ||
| GenBank mRNA | AK007725.1 | - | 4 | 1813 | ||
| AK084194.1 | - | 5 | 1875 | |||
| BC054756.1 | - | 4 | 1818 | 6411797 | ||