siRNA Details
| Gene Symbol: Zbtb8os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025970.3 | NP_080246.1 | 7 | 586 | ||
| GenBank mRNA | AK007377.1 | - | 7 | 576 | ||
| AK008007.1 | - | 7 | 587 | |||
| AK132279.1 | - | 597 | ||||
| BC026442.1 | - | 581 | ||||
| BC037657.1 | - | 556 | ||||
| BC046825.1 | - | 593 | ||||
| BC094629.1 | - | 7 | 617 | 3376726 | ||

Zbtb8os
Genomic Map