siRNA Details
| Gene Symbol: Naaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163687.1 | NP_001157159.1 | 8, 9 | 1035 | ||
| NM_025972.4 | NP_080248.2 | 8, 9 | 1041 | |||
| GenBank mRNA | AB162194.1 | - | 8, 9 | 1007 | ||
| AK008776.1 | - | 8, 9 | 1043 | |||
| AK017141.1 | - | 8, 9 | 1018 | |||
| AK133485.1 | - | 8, 9 | 1026 | |||
| AK133658.1 | - | 8, 9 | 1042 | |||
| AK146191.1 | - | 8, 9 | 1029 | |||
| BC004572.1 | - | 8, 9 | 1011 | 3586255 | ||

Naaa
Genomic Map