siRNA Details
| Gene Symbol: Bfar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025976.5 | NP_080252.1 | 4, 5 | 941 | ||
| GenBank mRNA | AK013874.1 | - | 4, 5 | 806 | ||
| BC027221.1 | - | 4, 5 | 782 | 4196466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bfar| Gene Symbol: Bfar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025976.5 | NP_080252.1 | 4, 5 | 941 | ||
| GenBank mRNA | AK013874.1 | - | 4, 5 | 806 | ||
| BC027221.1 | - | 4, 5 | 782 | 4196466 | ||