siRNA Details
| Gene Symbol: Acbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025988.2 | NP_080264.1 | 10 | 2033 | ||
| GenBank mRNA | AK008170.1 | - | 6 | 1014 | ||
| AK008207.1 | - | 4 | 1093 | |||
| AK008243.1 | - | 8 | 1582 | |||
| AK079604.1 | - | 9 | 1902 | |||
| AK148438.1 | - | 1529 | ||||
| AK165381.1 | - | 4 | 1170 | |||
| BC048371.1 | - | 9 | 1947 | 6396076 | ||

Acbd4
Genomic Map