siRNA Details
| Gene Symbol: Efhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025994.3 | NP_080270.2 | 1 | 322 | ||
| GenBank mRNA | AK007560.1 | - | 1 | 312 | ||
| AK011219.1 | - | 1 | 311 | |||
| AK048475.1 | - | 1 | 323 | |||
| AK150207.1 | - | 1 | 305 | |||
| AK159367.1 | - | 1 | 312 | |||
| AK167267.1 | - | 1 | 313 | |||
| AK169409.1 | - | 1 | 312 | |||
| AK195742.1 | - | 1 | 38 | |||
| BC125303.1 | - | 1 | 354 | 40129818 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001031648, BC098936.

Efhd2
Genomic Map