siRNA Details
| Gene Symbol: Utp11l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026031.3 | NP_080307.1 | 3 | 231 | ||
| GenBank mRNA | AK012550.1 | - | 3 | 232 | ||
| AK084149.1 | - | 1 | 570 | |||
| AK145266.1 | - | 3 | 264 | |||
| AK147113.1 | - | 3 | 262 | |||
| BC043139.1 | - | 3 | 211 | |||
| BC061074.1 | - | 3 | 260 | 6706808 | ||

Utp11l
Genomic Map