siRNA Details
| Gene Symbol: Uqcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026063.2 | NP_080339.1 | 1, 2 | 145 | ||
| GenBank mRNA | AK013521.1 | - | 1, 2 | 149 | ||
| AK019014.1 | - | 1, 2 | 146 | |||
| BC030629.1 | - | 1, 2 | 148 | 1054665 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uqcc2| Gene Symbol: Uqcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026063.2 | NP_080339.1 | 1, 2 | 145 | ||
| GenBank mRNA | AK013521.1 | - | 1, 2 | 149 | ||
| AK019014.1 | - | 1, 2 | 146 | |||
| BC030629.1 | - | 1, 2 | 148 | 1054665 | ||