siRNA Details
| Gene Symbol: Ccdc53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122960.1 | NP_001116432.1 | 2, 3 | 300 | ||
| NM_026070.3 | NP_080346.1 | 2, 3 | 300 | |||
| NM_027487.1 | NP_081763.1 | 2, 3 | 300 | |||
| GenBank mRNA | AK004206.1 | - | 2, 3 | 215 | ||
| AK009358.1 | - | 2, 3 | 196 | |||
| AK013835.1 | - | 2, 3 | 305 | |||
| AK017732.1 | - | 2, 3 | 228 | |||
| AK028599.1 | - | 1, 2 | 733 | |||
| AK158822.1 | - | 2, 3 | 301 | |||
| BC029256.1 | - | 2, 3 | 197 | 3661472 | ||
| CK795584.1 | - | 2, 3 | 252 | |||

Ccdc53
Genomic Map