siRNA Details
| Gene Symbol: Mrps24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026080.2 | NP_080356.1 | 3, 4 | 265 | ||
| GenBank mRNA | AK003117.1 | - | 3, 4 | 249 | ||
| AK007338.1 | - | 3, 4 | 249 | |||
| AK010590.1 | - | 3, 4 | 246 | |||
| AK012461.1 | - | 3, 4 | 254 | |||
| AK014099.1 | - | 3, 4 | 252 | |||
| BC029262.1 | - | 3, 4 | 239 | 4911699 | ||
| BY100364.1 | - | 3, 4 | 268 | |||

Mrps24
Genomic Map