siRNA Details
| Gene Symbol: Dock7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290636.1 | NP_001277565.1 | 47 | 6251 | ||
| NM_026082.5 | NP_080358.3 | 46 | 6161 | |||
| GenBank mRNA | AK013336.1 | - | 5 | 540 | ||
| AK014226.1 | - | 3 | 364 | |||
| AK122554.2 | - | 16 | 1895 | |||
| BC024823.1 | - | 17 | 2263 | |||
| BC024917.1 | - | 11 | 1416 | |||
| BC055032.1 | - | 6 | 657 | |||
| BC086672.1 | - | 27 | 3669 | 30647593 | ||
| BC094420.1 | - | 35 | 4452 | |||

Dock7
Genomic Map