siRNA Details
| Gene Symbol: Smarca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011416.2 | NP_035546.2 | 29 | 4395 | ||
| NM_026003.2 | NP_080279.1 | 5 | 544 | |||
| GenBank mRNA | AK009309.1 | - | 1 | 54 | ||
| AK011935.1 | - | 5 | 489 | |||
| BC022975.1 | - | 7 | 1021 | |||
| BC056199.1 | - | 19 | 2493 | |||
| BC075641.1 | - | 29 | 4377 | 30544092 | ||

Smarca2
Genomic Map