siRNA Details
| Gene Symbol: Ube2t | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278115.1 | NP_001265044.1 | 2 | 200 | ||
| NM_026024.3 | NP_080300.1 | 2 | 377 | |||
| GenBank mRNA | AK012565.1 | - | 2 | 336 | ||
| AK021213.1 | - | 2 | 366 | |||
| AK166098.1 | - | 2 | 347 | |||
| BC029213.1 | - | 2 | 326 | 3591557 | ||
| CK334733.1 | - | 2 | 191 | |||

Ube2t
Genomic Map