siRNA Details
| Gene Symbol: Glod4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026029.3 | NP_080305.2 | 3 | 252 | ||
| GenBank mRNA | AK006979.1 | - | 3 | 575 | ||
| AK007439.1 | - | 3 | 217 | |||
| AK010992.1 | - | 3 | 210 | |||
| AK012569.1 | - | 3 | 230 | |||
| AK013212.1 | - | 3 | 230 | |||
| AK138073.1 | - | 229 | ||||
| BC061012.1 | - | 3 | 244 | 6587019 | ||
| BY122171.1 | - | 3 | 252 | |||

Glod4
Genomic Map