siRNA Details
| Gene Symbol: TESK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318230.1 | NP_001305159.1 | 2, 3 | 685 | ||
| NM_006285.2 | NP_006276.2 | 2, 3 | 685 | |||
| GenBank mRNA | AF507907.1 | - | 1, 2 | 98 | ||
| AK298907.1 | - | 2, 3 | 223 | |||
| BC037561.1 | - | 2, 3 | 280 | |||
| BC038448.1 | - | 2, 3 | 659 | 5170888 | ||
| BC067130.1 | - | 2, 3 | 696 | 6153652 | ||
| D50863.1 | - | 2, 3 | 621 | |||

TESK1
Genomic Map