siRNA Details
| Gene Symbol: Ccdc53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122960.1 | NP_001116432.1 | 5 | 546 | ||
| NM_026070.3 | NP_080346.1 | 5 | 549 | |||
| NM_027487.1 | NP_081763.1 | 5 | 549 | |||
| GenBank mRNA | AK004206.1 | - | 5 | 464 | ||
| AK009358.1 | - | 5 | 445 | |||
| AK013835.1 | - | 5 | 554 | |||
| AK017732.1 | - | 5 | 477 | |||
| AK158822.1 | - | 5 | 550 | |||
| CK795584.1 | - | 5 | 498 | |||

Ccdc53
Genomic Map