siRNA Details
| Gene Symbol: 1700037H04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026091.2 | NP_080367.1 | 3, 4 | 336 | ||
| GenBank mRNA | AK003406.1 | - | 3, 4 | 337 | ||
| AK006622.1 | - | 3, 4 | 406 | |||
| AK178717.1 | - | 1, 2 | 84 | |||
| BC006041.1 | - | 2, 3 | 248 | 3497476 | ||

1700037H04Rik
Genomic Map