siRNA Details
| Gene Symbol: Tex43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026099.3 | NP_080375.2 | 3 | 349 | ||
| GenBank mRNA | AK006893.1 | - | 3 | 350 | ||
| AK018985.1 | - | 3 | 349 | |||
| BC099538.1 | - | 3 | 346 | 30916077 | ||
| FJ211594.1 | - | 3 | 333 | |||

Tex43
Genomic Map