siRNA Details
| Gene Symbol: Tctex1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163767.1 | NP_001157239.1 | 5 | 566 | ||
| NM_001163768.1 | NP_001157240.1 | 4 | 473 | |||
| NM_026100.3 | NP_080376.1 | 5 | 598 | |||
| GenBank mRNA | AK006804.1 | - | 5 | 567 | ||
| AK015315.1 | - | 5 | 601 | |||
| BB634690.1 | - | 4 | 485 | |||
| BC048635.1 | - | 5 | 538 | |||
| BC099549.1 | - | 5 | 572 | 30919464 | ||
| BC132398.1 | - | 5 | 516 | 40130675 | ||
| BC132400.1 | - | 5 | 516 | 40130677 | ||
| BC145187.1 | - | 5 | 516 | 9053738 | ||

Tctex1d1
Genomic Map