siRNA Details
| Gene Symbol: Efhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025994.3 | NP_080270.2 | 2 | 487 | ||
| GenBank mRNA | AK007560.1 | - | 2 | 477 | ||
| AK011219.1 | - | 2 | 476 | |||
| AK048475.1 | - | 2 | 488 | |||
| AK150207.1 | - | 2 | 470 | |||
| AK159367.1 | - | 2 | 477 | |||
| AK167267.1 | - | 2 | 478 | |||
| AK169409.1 | - | 2 | 477 | |||
| AK195742.1 | - | 2 | 203 | |||
| BC125303.1 | - | 2 | 519 | 40129818 | ||

Efhd2
Genomic Map