siRNA Details
| Gene Symbol: Dram2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025582.2 | NP_001020753.1 | 7 | 947 | ||
| NM_001286986.1 | NP_001273915.1 | 9 | 1205 | |||
| NM_001286987.1 | NP_001273916.1 | 8 | 1083 | |||
| NM_026013.3 | NP_080289.1 | 9 | 1208 | |||
| GenBank mRNA | AK008532.1 | - | 7 | 871 | ||
| AK009940.1 | - | 9 | 1145 | |||
| AK012044.1 | - | 9 | 1125 | |||
| AK015888.1 | - | 9 | 1171 | |||
| AK052824.1 | - | 8 | 1017 | |||
| AK172455.1 | - | 9 | 1175 | |||
| BC024458.1 | - | 7 | 873 | 4975302 | ||

Dram2
Genomic Map