siRNA Details
| Gene Symbol: Fam69a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026062.4 | NP_080338.1 | 4, 5 | 557 | ||
| GenBank mRNA | AK008504.1 | - | 4, 5 | 500 | ||
| AK013580.1 | - | 4, 5 | 521 | |||
| AK038811.1 | - | 4, 5 | 514 | |||
| AK048979.1 | - | 4, 5 | 514 | |||
| AK050305.1 | - | 4, 5 | 514 | |||
| AK053273.1 | - | 3, 4 | 362 | |||
| BC055842.1 | - | 4, 5 | 489 | |||
| BC061013.1 | - | 4, 5 | 509 | |||
| BC095935.1 | - | 4, 5 | 491 | |||
| BC116683.1 | - | 4, 5 | 502 | 40106565 | ||

Fam69a
Genomic Map