siRNA Details
| Gene Symbol: Myl12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026064.2 | NP_080340.2 | 4 | 1654 | ||
| GenBank mRNA | AK013776.1 | - | 4 | 1341 | ||
| AK018348.1 | - | 4 | 1309 | |||
| BC010265.1 | - | 3 | 1239 | 3588394 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myl12a| Gene Symbol: Myl12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026064.2 | NP_080340.2 | 4 | 1654 | ||
| GenBank mRNA | AK013776.1 | - | 4 | 1341 | ||
| AK018348.1 | - | 4 | 1309 | |||
| BC010265.1 | - | 3 | 1239 | 3588394 | ||