siRNA Details
| Gene Symbol: Zc3h13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026083.2 | NP_080359.2 | 10 | 2121 | ||
| GenBank mRNA | AK014200.1 | - | 10 | 1902 | ||
| AK078225.1 | - | 10 | 2112 | |||
| BC038882.1 | - | 3 | 931 | |||
| BC094575.1 | - | 2 | 725 | |||
| BC138264.1 | - | 10 | 2070 | 8861283 | ||

Zc3h13
Genomic Map