siRNA Details
| Gene Symbol: 1110008F13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026124.3 | NP_080400.1 | 2 | 389 | ||
| GenBank mRNA | AK002452.1 | - | 2 | 347 | ||
| AK003568.1 | - | 2 | 384 | |||
| AK031002.1 | - | 2 | 368 | |||
| BC019445.1 | - | 2 | 327 | 5125733 | ||

1110008F13Rik
Genomic Map