siRNA Details
| Gene Symbol: Klhl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290476.1 | NP_001277405.1 | 2 | 295 | ||
| NM_026167.4 | NP_080443.1 | 4 | 314 | |||
| GenBank mRNA | AK004677.1 | - | 4 | 286 | ||
| AK122491.1 | - | 2 | 295 | |||
| BY136647.1 | - | 4 | 314 | |||

Klhl13
Genomic Map