siRNA Details
| Gene Symbol: Sf3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026175.5 | NP_080451.4 | 13, 14 | 2235 | ||
| GenBank mRNA | AK004763.1 | - | 13, 14 | 2218 | ||
| AK008075.1 | - | 2, 3 | 262 | |||
| AK028829.1 | - | 13, 14 | 2187 | |||
| AK029095.1 | - | 13, 14 | 2205 | |||
| AK030115.1 | - | 5, 6 | 761 | |||
| AK030223.1 | - | 13, 14 | 2222 | |||
| AK134178.1 | - | 13, 14 | 2213 | |||
| AK160061.1 | - | 13, 14 | 2218 | |||
| BC010727.1 | - | 4, 5 | 690 | |||
| BC029753.1 | - | 13, 14 | 2199 | 2651773 | ||

Sf3a1
Genomic Map