siRNA Details
| Gene Symbol: TESK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318230.1 | NP_001305159.1 | 5, 6 | 972 | ||
| NM_006285.2 | NP_006276.2 | 6, 7 | 1055 | |||
| GenBank mRNA | AF507907.1 | - | 5, 6 | 468 | ||
| AK298907.1 | - | 6, 7 | 593 | |||
| BC037561.1 | - | 6, 7 | 650 | |||
| BC038448.1 | - | 6, 7 | 1029 | 5170888 | ||
| BC067130.1 | - | 6, 7 | 1066 | 6153652 | ||
| D50863.1 | - | 6, 7 | 991 | |||

TESK1
Genomic Map