siRNA Details
| Gene Symbol: SLC26A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000112.3 | NP_000103.2 | 3 | 2504 | ||
| GenBank mRNA | AK290358.1 | - | 3 | 2327 | ||
| BC059390.1 | - | 3 | 2358 | 30341764 | ||
| U14528.1 | - | 2 | 2263 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC26A2| Gene Symbol: SLC26A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000112.3 | NP_000103.2 | 3 | 2504 | ||
| GenBank mRNA | AK290358.1 | - | 3 | 2327 | ||
| BC059390.1 | - | 3 | 2358 | 30341764 | ||
| U14528.1 | - | 2 | 2263 | |||