siRNA Details
| Gene Symbol: Tvp23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026210.4 | NP_080486.1 | 5 | 465 | ||
| GenBank mRNA | AK007707.1 | - | 5 | 381 | ||
| AK017572.1 | - | 5 | 380 | |||
| AK050027.1 | - | 5 | 501 | |||
| AK077277.1 | - | 5 | 467 | |||
| AK150936.1 | - | 5 | 457 | |||
| BC115504.1 | - | 5 | 445 | 40101013 | ||
| BC115505.1 | - | 5 | 445 | 40101015 | ||

Tvp23b
Genomic Map