siRNA Details
Gene Symbol: Mfap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081975.3 | NP_001075444.1 | 1, 2 | 198 | ||
GenBank mRNA | AK011317.1 | - | 1, 2 | 196 | ||
AK018514.1 | - | 1, 2 | 199 | |||
AK088946.1 | - | 205 | ||||
AK169029.1 | - | 206 | ||||
BC005728.1 | - | 1, 2 | 159 | 3595848 |
Gene Symbol: Mfap1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026220.4 | NP_080496.1 | 1, 2 | 202 | ||
GenBank mRNA | AK012688.1 | - | 1, 2 | 188 | ||
AK014483.1 | - | 1, 2 | 195 | |||
AK032586.1 | - | 1, 2 | 203 | |||
AK050561.1 | - | 1, 2 | 203 | |||
AK077604.1 | - | 1, 2 | 196 | |||
AK088766.1 | - | 1, 2 | 199 | |||
AK161000.1 | - | 1, 2 | 203 | |||
AK168801.1 | - | 1, 2 | 208 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
