siRNA Details
| Gene Symbol: Zfp606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026112.4 | NP_080388.2 | 6 | 841 | ||
| GenBank mRNA | AK010575.1 | - | 6 | 836 | ||
| AK021179.1 | - | 6 | 790 | |||
| AK044645.1 | - | 6 | 809 | |||
| AK077352.1 | - | 2 | 265 | |||
| AK139269.1 | - | 6 | 842 | |||
| AK143200.1 | - | 6 | 806 | |||
| AK173278.1 | - | 7 | 847 | |||
| BC052747.1 | - | 6 | 760 | 6837952 | ||

Zfp606
Genomic Map