siRNA Details
| Gene Symbol: 2410127L17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026120.4 | NP_080396.2 | 2 | 561 | ||
| GenBank mRNA | AI509964.1 | - | 2 | 347 | ||
| AK010784.1 | - | 2 | 446 | |||
| BB656580.1 | - | 2 | 577 | |||
| BC019559.1 | - | 2 | 751 | |||
| BC043335.1 | - | 2 | 515 | 5030742 | ||
| BC092281.1 | - | 2 | 393 | 4950426 | ||

2410127L17Rik
Genomic Map