siRNA Details
| Gene Symbol: Klhl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290476.1 | NP_001277405.1 | 2 | 364 | ||
| NM_026167.4 | NP_080443.1 | 4 | 383 | |||
| GenBank mRNA | AK004677.1 | - | 4 | 355 | ||
| AK122491.1 | - | 2 | 364 | |||
| AK191256.1 | - | 1 | 90 | |||
| BC100393.1 | - | 3 | 348 | 30737426 | ||

Klhl13
Genomic Map