siRNA Details
| Gene Symbol: Gpalpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026177.3 | NP_080453.2 | 2 | 297 | ||
| GenBank mRNA | AK004707.1 | - | 2 | 277 | ||
| AK016681.1 | - | 2 | 307 | |||
| AK082912.1 | - | 2 | 278 | |||
| AK173238.1 | - | 2 | 242 | |||
| BC029731.1 | - | 2 | 258 | 4946215 | ||

Gpalpp1
Genomic Map