siRNA Details
| Gene Symbol: Slc47a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026183.5 | NP_080459.2 | 12 | 1110 | ||
| GenBank mRNA | AK004994.2 | - | 13 | 1466 | ||
| AK009038.1 | - | 7 | 730 | |||
| BC031436.1 | - | 12 | 1094 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc47a1| Gene Symbol: Slc47a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026183.5 | NP_080459.2 | 12 | 1110 | ||
| GenBank mRNA | AK004994.2 | - | 13 | 1466 | ||
| AK009038.1 | - | 7 | 730 | |||
| BC031436.1 | - | 12 | 1094 | |||