siRNA Details
| Gene Symbol: Ap4b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163552.1 | NP_001157024.1 | 11 | 2032 | ||
| NM_001163553.1 | NP_001157025.1 | 9 | 2161 | |||
| NM_026193.2 | NP_080469.2 | 10 | 2245 | |||
| GenBank mRNA | AK003021.1 | - | 2 | 300 | ||
| AK007733.1 | - | 10 | 2134 | |||

Ap4b1
Genomic Map